Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025631.3 | NP_079907.2 | 5 - 6 | 566 | 89 | |
GenBank mRNA | AK009346.1 | - | 5 - 6 | 539 | 89 | |
AK009504.1 | - | 5 - 6 | 541 | 89 | ||
AK009619.1 | - | 5 - 6 | 539 | 89 | ||
AK009754.1 | - | 1 - 2 | 142 | 89 | ||
AK009776.1 | - | 5 - 6 | 523 | 89 | ||
AK009794.1 | - | 5 - 6 | 537 | 89 | ||
AK009871.1 | - | 5 - 6 | 541 | 89 | ||
AK010130.1 | - | 5 - 6 | 523 | 89 | ||
AK010154.1 | - | 5 - 6 | 537 | 89 | ||
AK010157.1 | - | 5 - 6 | 567 | 89 | ||
AK010215.1 | - | 5 - 6 | 537 | 89 | ||
AK028156.1 | - | 5 - 6 | 537 | 89 | ||
AK028158.1 | - | 5 - 6 | 540 | 89 | ||
AK075874.1 | - | 5 - 6 | 537 | 89 | ||
AK160516.1 | - | 5 - 6 | 537 | 89 | ||
BC116681.1 | - | 5 - 6 | 521 | 40106489 | 89 | |
BC116727.1 | - | 5 - 6 | 521 | 40106498 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |