Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025631.3 | NP_079907.2 | 9 - 10 | 963 | 63 | |
GenBank mRNA | AK009346.1 | - | 9 - 10 | 935 | 63 | |
AK009619.1 | - | 9 - 10 | 936 | 63 | ||
AK009754.1 | - | 5 - 6 | 539 | 63 | ||
AK009776.1 | - | 9 - 10 | 921 | 63 | ||
AK009871.1 | - | 9 - 10 | 932 | 63 | ||
AK010154.1 | - | 9 - 10 | 931 | 63 | ||
AK010157.1 | - | 9 - 10 | 962 | 63 | ||
AK010215.1 | - | 9 - 10 | 932 | 63 | ||
AK028156.1 | - | 9 - 10 | 934 | 63 | ||
AK028158.1 | - | 9 - 10 | 937 | 63 | ||
AK075874.1 | - | 9 - 10 | 934 | 63 | ||
AK160516.1 | - | 9 - 10 | 934 | 63 | ||
BC116681.1 | - | 9 - 10 | 918 | 40106489 | 63 | |
BC116727.1 | - | 9 - 10 | 918 | 40106498 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |