Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008916.2 | NP_032942.1 | 2 - 3 | 290 | 84 | |
GenBank mRNA | AF483522.1 | - | 2 - 3 | 210 | 84 | |
AF483523.1 | - | 2 - 3 | 210 | 84 | ||
AK052817.1 | - | 2 - 3 | 282 | 84 | ||
AK054436.1 | - | 2 - 3 | 309 | 84 | ||
AK078104.1 | - | 2 - 3 | 301 | 84 | ||
AK085743.1 | - | 2 - 3 | 274 | 84 | ||
AK164290.1 | - | 2 - 3 | 279 | 84 | ||
BC014725.1 | - | 2 - 3 | 220 | 3969089 | 84 | |
BC066112.1 | - | 2 - 3 | 290 | 30361046 | 84 | |
U96724.1 | - | 2 - 3 | 321 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |