Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rilp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029938.2 | NP_001025109.1 | 1 - 2 | 279 | 87 | |
GenBank mRNA | AK139016.1 | - | 1 - 2 | 271 | 87 | |
BC116750.1 | - | 1 - 2 | 263 | 40126069 | 87 | |
BC119166.1 | - | 1 - 2 | 263 | 8733915 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |