Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200020904.mm.1 |
NM_011611.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020904.mm.1 |
NM_011611.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020904.mm.1 |
NM_170702.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020904.mm.1 |
NM_170702.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020904.mm.1 |
NM_170703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020904.mm.1 |
NM_170703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020904.mm.1 |
NM_170704.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020904.mm.1 |
NM_170704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020904.mm.1 |
NR_027852.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020904.mm.1 |
NR_027852.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020904.mm.1 |
TC0200002544.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200020901.mm.1 |
NM_011611.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020901.mm.1 |
NM_011611.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020901.mm.1 |
NM_170702.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020901.mm.1 |
NM_170702.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020901.mm.1 |
NM_170703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020901.mm.1 |
NM_170703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020901.mm.1 |
NM_170704.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020901.mm.1 |
NM_170704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020901.mm.1 |
NR_027852.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020901.mm.1 |
NR_027852.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020901.mm.1 |
TC0200002544.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200020902.mm.1 |
NM_011611.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020902.mm.1 |
NM_011611.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020902.mm.1 |
NM_170702.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020902.mm.1 |
NM_170702.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020902.mm.1 |
NM_170703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020902.mm.1 |
NM_170703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020902.mm.1 |
NM_170704.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020902.mm.1 |
NM_170704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020902.mm.1 |
NR_027852.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020902.mm.1 |
NR_027852.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020902.mm.1 |
TC0200002544.mm.1 |
MTA TC |