Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011611.2 | NP_035741.2 | 6 - 7 | 637 | 88 | |
NM_170704.2 | NP_733805.1 | 6 - 7 | 637 | 88 | ||
NR_027852.1 | 6 - 7 | 637 | 88 | |||
GenBank mRNA | AJ401388.1 | - | 6 - 7 | 571 | 88 | |
AJ401389.1 | - | 6 - 7 | 571 | 88 | ||
AK089861.1 | - | 590 | 88 | |||
AK150959.1 | - | 6 - 7 | 594 | 88 | ||
AK151458.1 | - | 6 - 7 | 594 | 88 | ||
AK152716.1 | - | 6 - 7 | 595 | 88 | ||
AK152756.1 | - | 6 - 7 | 593 | 88 | ||
AK152942.1 | - | 6 - 7 | 601 | 88 | ||
AK156644.1 | - | 6 - 7 | 605 | 88 | ||
AK161978.1 | - | 6 - 7 | 594 | 88 | ||
AK162305.1 | - | 6 - 7 | 647 | 88 | ||
BC029254.1 | - | 6 - 7 | 595 | 4018221 | 88 | |
BY743050.1 | - | 6 - 7 | 593 | 88 | ||
CT010343.1 | - | 6 - 7 | 565 | 88 | ||
M83312.1 | - | 6 - 7 | 573 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |