Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crispld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030209.3 | NP_084485.1 | 2 - 3 | 449 | 79 | |
GenBank mRNA | AK019034.1 | - | 2 - 3 | 445 | 79 | |
AK033858.1 | - | 2 - 3 | 410 | 79 | ||
AK132171.1 | - | 2 - 3 | 392 | 79 | ||
AK149909.1 | - | 2 - 3 | 406 | 79 | ||
AK149925.1 | - | 2 - 3 | 406 | 79 | ||
AK150180.1 | - | 2 - 3 | 406 | 79 | ||
AK154308.1 | - | 2 - 3 | 449 | 79 | ||
BC090642.1 | - | 2 - 3 | 443 | 30653388 | 79 | |
CN527154.1 | - | 2 - 3 | 443 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |