Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133924.3 | NP_598685.2 | 1 - 2 | 305 | 120 | |
GenBank mRNA | AK017516.1 | - | 1 - 2 | 310 | 120 | |
AK141663.1 | - | 1 - 2 | 306 | 120 | ||
AK168473.1 | - | 1 - 2 | 292 | 120 | ||
BC117542.1 | - | 1 - 2 | 514 | 40105600 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |