Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018771.3 | NP_061241.1 | 4 - 5 | 744 | 70 | |
GenBank mRNA | AF061263.1 | - | 3 - 4 | 678 | 70 | |
AF104358.1 | - | 4 - 5 | 722 | 70 | ||
AK002492.1 | - | 4 - 5 | 725 | 70 | ||
AK086506.1 | - | 4 - 5 | 745 | 70 | ||
BC003490.1 | - | 4 - 5 | 744 | 2811749 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |