Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800019045.mm.1 |
NM_145599.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019045.mm.1 |
NM_145599.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019045.mm.1 |
TC0800002503.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019046.mm.1 |
NM_145599.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019046.mm.1 |
NM_145599.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019046.mm.1 |
TC0800002503.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019048.mm.1 |
NM_145599.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019048.mm.1 |
NM_145599.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019048.mm.1 |
TC0800002503.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019044.mm.1 |
NM_145599.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019044.mm.1 |
NM_145599.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019044.mm.1 |
TC0800002503.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019047.mm.1 |
NM_145599.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019047.mm.1 |
NM_145599.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019047.mm.1 |
TC0800002503.mm.1 |
MTA TC |