Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172757.3 | NP_766345.3 | 4 - 5 | 643 | 94 | |
GenBank mRNA | AK047782.1 | - | 4 - 5 | 674 | 94 | |
AK052635.1 | - | 4 - 5 | 670 | 94 | ||
AK136424.1 | - | 4 - 5 | 653 | 94 | ||
AK163715.1 | - | 4 - 5 | 630 | 94 | ||
AK165541.1 | - | 4 - 5 | 538 | 93 | ||
BC065060.1 | - | 4 - 5 | 643 | 6402217 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |