Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145605.2 | NP_663580.2 | 3 - 4 | 361 | 91 | |
GenBank mRNA | AK089954.1 | - | 3 - 4 | 324 | 91 | |
AK161674.1 | - | 3 - 4 | 337 | 91 | ||
BC012312.1 | - | 3 - 4 | 361 | 2631632 | 91 | |
BC023738.1 | - | 3 - 4 | 332 | 5352770 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |