Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133765.4 | NP_598526.3 | 2 - 3 | 421 | 63 | |
GenBank mRNA | AB041586.1 | - | 2 - 3 | 270 | 63 | |
AK140412.1 | - | 2 - 3 | 422 | 63 | ||
AK163641.1 | - | 2 - 3 | 390 | 63 | ||
BC026929.1 | - | 2 - 3 | 406 | 4194857 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |