Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172945.2 | NP_766533.2 | 9 - 10 | 1096 | 82 | |
GenBank mRNA | BC056928.1 | - | 4 - 5 | 325 | 82 | |
BC071189.1 | - | 3 - 4 | 182 | 82 | ||
BC098207.1 | - | 2 - 3 | 135 | 82 | ||
BC151158.1 | - | 9 - 10 | 1074 | 9088046 | 82 | |
BC172028.1 | - | 9 - 10 | 1075 | 82 | ||
CD578820.1 | - | 3 - 4 | 194 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |