Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Milr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033435.3 | NP_001028607.1 | 1 - 2 | 163 | 89 | |
NM_001271372.1 | NP_001258301.1 | 1 - 2 | 163 | 89 | ||
NM_001271373.1 | NP_001258302.1 | 1 - 2 | 163 | 89 | ||
NM_001271374.1 | NP_001258303.1 | 1 - 2 | 163 | 89 | ||
NM_001271375.1 | NP_001258304.1 | 1 - 2 | 163 | 89 | ||
NR_073165.1 | 1 - 2 | 163 | 89 | |||
GenBank mRNA | AB542953.1 | - | 1 - 2 | 121 | 89 | |
AK136488.1 | - | 1 - 2 | 123 | 89 | ||
AK137500.1 | - | 1 - 2 | 122 | 89 | ||
AK151814.1 | - | 1 - 2 | 121 | 89 | ||
AK153506.1 | - | 1 - 2 | 71 | 89 | ||
AK169925.1 | - | 1 - 2 | 126 | 89 | ||
AK170947.1 | - | 1 - 2 | 122 | 89 | ||
AK171381.1 | - | 1 - 2 | 122 | 89 | ||
BC139224.1 | - | 1 - 2 | 149 | 8862246 | 89 | |
BC139225.1 | - | 1 - 2 | 149 | 8862247 | 89 | |
BC145580.1 | - | 1 - 2 | 149 | 9054146 | 89 | |
BC145582.1 | - | 1 - 2 | 149 | 9054148 | 89 | |
BY197245.1 | - | 1 - 2 | 167 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |