Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145896.1 | NP_001139368.1 | 3 - 4 | 578 | 116 | |
NM_001145897.1 | NP_001139369.1 | 3 - 4 | 522 | 116 | ||
NM_198671.2 | NP_941073.2 | 3 - 4 | 710 | 116 | ||
GenBank mRNA | AK033263.1 | - | 3 - 4 | 524 | 116 | |
AK042915.1 | - | 3 - 4 | 526 | 116 | ||
AK154829.1 | - | 3 - 4 | 557 | 116 | ||
BC043094.1 | - | 3 - 4 | 710 | 6405464 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |