Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008522.3 | NP_032548.2 | 10 - 11 | 1330 | 74 | |
GenBank mRNA | AK036491.1 | - | 10 - 11 | 1331 | 74 | |
AK144556.1 | - | 10 - 11 | 1331 | 74 | ||
AK150799.1 | - | 10 - 11 | 1331 | 74 | ||
AK151822.1 | - | 10 - 11 | 1331 | 74 | ||
AK164781.1 | - | 10 - 11 | 1332 | 74 | ||
BC006904.1 | - | 10 - 11 | 1338 | 3600033 | 74 | |
BC008530.1 | - | 10 - 11 | 1311 | 74 | ||
BC009662.1 | - | 6 - 7 | 761 | 74 | ||
CT010339.1 | - | 10 - 11 | 1296 | 74 | ||
D88510.1 | - | 10 - 11 | 1330 | 74 | ||
FJ538998.1 | - | 11 - 12 | 1296 | 74 | ||
J03298.1 | - | 10 - 11 | 1299 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |