Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081379.2 | NP_001074848.2 | 5 - 6 | 857 | 62 | |
NR_037865.1 | 6 - 7 | 954 | 62 | |||
GenBank mRNA | BC005576.1 | - | 3 - 4 | 454 | 62 | |
BC158043.1 | - | 4 - 5 | 522 | 9088159 | 62 | |
BC172141.1 | - | 5 - 6 | 619 | 9054811 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |