Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002764.1 | NP_001002764.1 | 6 - 7 | 2379 | 73 | |
GenBank mRNA | AK129203.1 | - | 5 - 6 | 998 | 73 | |
BC037179.1 | - | 5 - 6 | 731 | 73 | ||
BC047279.1 | - | 6 - 7 | 2391 | 73 | ||
BC066040.1 | - | 6 - 7 | 2379 | 6410245 | 73 | |
BC077715.1 | - | 6 - 7 | 2342 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |