Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138950.2 | NP_620400.2 | 3 - 4 | 4225 | 82 | |
GenBank mRNA | AK131146.1 | - | 3 - 4 | 825 | 82 | |
AK143628.1 | - | 2 - 3 | 313 | 82 | ||
AK160585.1 | - | 2 - 3 | 267 | 82 | ||
AK172019.1 | - | 3 - 4 | 591 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |