Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002929.4 | NP_001002929.3 | 10 - 11 | 1076 | 61 | |
GenBank mRNA | AF498263.1 | - | 10 - 11 | 990 | 61 | |
AK017632.1 | - | 10 - 11 | 1041 | 61 | ||
AK196154.1 | - | 1 - 2 | 116 | 61 | ||
BC079856.1 | - | 10 - 11 | 1051 | 6403446 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |