Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002929.4 | NP_001002929.3 | 12 - 13 | 1256 | 78 | |
GenBank mRNA | AF498263.1 | - | 12 - 13 | 1170 | 78 | |
AK017632.1 | - | 12 - 13 | 1221 | 78 | ||
BC079856.1 | - | 12 - 13 | 1231 | 6403446 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |