Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fanca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016925.3 | NP_058621.2 | 17 - 18 | 1642 | 88 | |
GenBank mRNA | AF178934.1 | - | 17 - 18 | 1638 | 88 | |
AF208116.1 | - | 17 - 18 | 1644 | 88 | ||
AF208117.1 | - | 14 - 15 | 1325 | 88 | ||
AF208118.1 | - | 17 - 18 | 1644 | 88 | ||
AK084990.1 | - | 16 - 17 | 1750 | 88 | ||
BC150705.1 | - | 17 - 18 | 1640 | 9087616 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |