Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fanca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016925.3 | NP_058621.2 | 21 - 22 | 1913 | 65 | |
GenBank mRNA | AF178934.1 | - | 21 - 22 | 1909 | 65 | |
AF208116.1 | - | 21 - 22 | 1915 | 65 | ||
AF208117.1 | - | 18 - 19 | 1596 | 65 | ||
AF208118.1 | - | 21 - 22 | 1915 | 65 | ||
AF208120.1 | - | 1 - 2 | 78 | 65 | ||
AK084990.1 | - | 20 - 21 | 2021 | 65 | ||
AK218957.1 | - | 1 - 2 | 39 | 65 | ||
BC150705.1 | - | 21 - 22 | 1911 | 9087616 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |