Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172758.4 | NP_766346.1 | 8 - 9 | 1126 | 65 | |
GenBank mRNA | AK052548.1 | - | 8 - 9 | 1127 | 65 | |
AK171137.1 | - | 8 - 9 | 1134 | 65 | ||
AK194183.1 | - | 3 - 4 | 242 | 65 | ||
BC031853.1 | - | 6 - 7 | 1001 | 4009949 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |