Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164497.1 | NP_001157969.1 | 10 - 11 | 1641 | 151 | |
NM_001164498.1 | NP_001157970.1 | 10 - 11 | 1615 | 151 | ||
NM_001164499.1 | NP_001157971.1 | 9 - 10 | 1512 | 151 | ||
GenBank mRNA | AK077553.1 | - | 10 - 11 | 1643 | 151 | |
BC059053.1 | - | 10 - 11 | 1613 | 151 | ||
BC080314.1 | - | 9 - 10 | 1216 | 151 | ||
BC144796.1 | - | 10 - 11 | 1599 | 9053344 | 151 | |
BC144797.1 | - | 9 - 10 | 1470 | 9053345 | 151 | |
BC145737.1 | - | 10 - 11 | 1599 | 40131040 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |