Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134141.4 | NP_598902.1 | 1 - 2 | 89 | 77 | |
GenBank mRNA | AK076116.1 | - | 1 - 2 | 45 | 77 | |
AK079615.1 | - | 1 - 2 | 51 | 77 | ||
AK136821.1 | - | 1 - 2 | 82 | 78 | ||
AK146282.1 | - | 1 - 2 | 37 | 77 | ||
AK163705.1 | - | 1 - 2 | 68 | 77 | ||
AK168252.1 | - | 1 - 2 | 39 | 77 | ||
BC021864.1 | - | 1 - 2 | 29 | 5123958 | 77 | |
BY283649.1 | - | 1 - 2 | 108 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |