Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182959.3 | NP_892004.1 | 7 - 8 | 1291 | 111 | |
GenBank mRNA | AF510321.1 | - | 7 - 8 | 962 | 111 | |
AK082743.1 | - | 7 - 8 | 1039 | 111 | ||
AK149687.1 | - | 5 - 6 | 694 | 111 | ||
BC042593.1 | - | 7 - 8 | 1306 | 5367275 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |