Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps15l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122832.1 | NP_001116304.1 | 12 - 13 | 1223 | 96 | |
NM_001289859.1 | NP_001276788.1 | 12 - 13 | 1223 | 96 | ||
NM_007944.3 | NP_031970.2 | 12 - 13 | 1223 | 96 | ||
GenBank mRNA | AK036662.1 | - | 12 - 13 | 1189 | 96 | |
AK036728.1 | - | 12 - 13 | 1201 | 96 | ||
AK146781.1 | - | 12 - 13 | 1201 | 96 | ||
AK152602.1 | - | 12 - 13 | 1201 | 96 | ||
BC015259.1 | - | 12 - 13 | 1204 | 4238768 | 96 | |
U29156.1 | - | 12 - 13 | 1210 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |