Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007440.4 | NP_031466.2 | 1 - 2 | 207 | 68 | |
GenBank mRNA | AK036898.1 | - | 1 - 2 | 205 | 68 | |
AK079232.1 | - | 1 - 2 | 208 | 68 | ||
AK087724.1 | - | 1 - 2 | 208 | 68 | ||
AK172216.1 | - | 1 - 2 | 205 | 68 | ||
AK172488.1 | - | 1 - 2 | 208 | 68 | ||
BC152329.1 | - | 1 - 2 | 212 | 40141477 | 68 | |
BC152403.1 | - | 1 - 2 | 212 | 40141482 | 68 | |
S80446.1 | - | 1 - 2 | 219 | 68 | ||
U04334.1 | - | 1 - 2 | 138 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |