Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190376.1 | NP_001177305.1 | 21 - 22 | 3759 | 109 | |
NM_001190378.1 | NP_001177307.1 | 20 - 21 | 3738 | 109 | ||
NM_001190379.1 | NP_001177308.1 | 19 - 20 | 3666 | 109 | ||
NM_178116.4 | NP_835217.1 | 21 - 22 | 3774 | 109 | ||
GenBank mRNA | AK122400.1 | - | 19 - 20 | 3415 | 109 | |
AK150186.1 | - | 20 - 21 | 3593 | 110 | ||
AK169118.1 | - | 19 - 20 | 3505 | 109 | ||
BC023962.1 | - | 15 - 16 | 3552 | 109 | ||
BC027039.1 | - | 10 - 11 | 1483 | 109 | ||
BC038079.1 | - | 3 - 4 | 365 | 109 | ||
BC049133.1 | - | 21 - 22 | 3603 | 6827800 | 109 | |
BC056395.1 | - | 21 - 22 | 3588 | 6852349 | 109 | |
BC059065.1 | - | 19 - 20 | 3533 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |