Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177776.3 | NP_808444.1 | 2 - 3 | 560 | 103 | |
GenBank mRNA | AK051585.1 | - | 2 - 3 | 559 | 103 | |
AK085988.1 | - | 2 - 3 | 104 | |||
BC037993.1 | - | 2 - 3 | 558 | 5064159 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |