Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177776.3 | NP_808444.1 | 6 - 7 | 1171 | 77 | |
GenBank mRNA | AK051585.1 | - | 6 - 7 | 1171 | 77 | |
BC037993.1 | - | 6 - 7 | 1169 | 5064159 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |