Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK144538.1 | - | 5 - 6 | 469 | 149 | |
AK144683.1 | - | 3 - 4 | 223 | 149 | ||
AK158497.1 | - | 4 - 5 | 531 | 149 | ||
AK166136.1 | - | 5 - 6 | 464 | 149 | ||
DQ832277.1 | - | 4 - 5 | 232 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |