Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubtf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004961.1 | - | 1 - 2 | 116 | 114 | |
AK128947.1 | - | 1 - 2 | 143 | 114 | ||
BC004676.1 | - | 1 - 2 | 249 | 114 | ||
BY705170.1 | - | 1 - 2 | 116 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |