Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161329.1 | NP_001154801.1 | 2 - 3 | 378 | 123 | |
NM_133702.2 | NP_598463.2 | 2 - 3 | 378 | 123 | ||
GenBank mRNA | AK005123.1 | - | 2 - 3 | 268 | 123 | |
AK038696.1 | - | 2 - 3 | 269 | 123 | ||
AK076704.1 | - | 2 - 3 | 379 | 123 | ||
AK078524.1 | - | 2 - 3 | 269 | 123 | ||
BC024874.1 | - | 2 - 3 | 257 | 3984027 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |