Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161329.1 | NP_001154801.1 | 5 - 6 | 645 | 122 | |
NM_133702.2 | NP_598463.2 | 5 - 6 | 645 | 122 | ||
GenBank mRNA | AK005123.1 | - | 5 - 6 | 535 | 122 | |
AK038696.1 | - | 5 - 6 | 536 | 122 | ||
AK076704.1 | - | 5 - 6 | 646 | 122 | ||
AK078524.1 | - | 5 - 6 | 536 | 122 | ||
AK157743.1 | - | 5 - 6 | 585 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |