Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200018526.mm.1 |
NM_144890.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018526.mm.1 |
NM_144890.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018526.mm.1 |
TC0200002338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018528.mm.1 |
NM_144890.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018528.mm.1 |
NM_144890.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018528.mm.1 |
TC0200002338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018527.mm.1 |
NM_144890.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018527.mm.1 |
NM_144890.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018527.mm.1 |
TC0200002338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018524.mm.1 |
NM_144890.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018524.mm.1 |
NM_144890.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018524.mm.1 |
TC0200002338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018525.mm.1 |
NM_144890.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018525.mm.1 |
NM_144890.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018525.mm.1 |
TC0200002338.mm.1 |
MTA TC |