Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008682.2 | NP_032708.2 | 3 - 4 | 332 | 90 | |
GenBank mRNA | AK010591.1 | - | 3 - 4 | 320 | 90 | |
AK048393.1 | - | 3 - 4 | 284 | 90 | ||
AK085355.1 | - | 3 - 4 | 295 | 90 | ||
AK090040.1 | - | 3 - 4 | 277 | 90 | ||
AK153746.1 | - | 3 - 4 | 334 | 90 | ||
BC052430.1 | - | 3 - 4 | 301 | 6836583 | 90 | |
BC066870.1 | - | 3 - 4 | 269 | 30476383 | 90 | |
D10712.1 | - | 3 - 4 | 274 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |