Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emilin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291145.1 | NP_001278074.1 | 3 - 4 | 683 | 97 | |
GenBank mRNA | BC002161.1 | - | 2 - 3 | 311 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |