Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Banp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110100.2 | NP_001103570.1 | 5 - 6 | 705 | 79 | |
NM_016812.4 | NP_058092.2 | 5 - 6 | 705 | 79 | ||
GenBank mRNA | AF235503.1 | - | 5 - 6 | 616 | 79 | |
AK178331.1 | - | 2 - 3 | 281 | 79 | ||
AK203167.1 | - | 2 - 3 | 298 | 79 | ||
AK208082.1 | - | 2 - 3 | 281 | 79 | ||
BC021650.1 | - | 5 - 6 | 623 | 3660187 | 79 | |
BC022168.1 | - | 5 - 6 | 623 | 4189797 | 79 | |
BC062641.1 | - | 5 - 6 | 650 | 5718330 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |