Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800010215.mm.1 |
NM_001164472.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010215.mm.1 |
NM_001164472.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010215.mm.1 |
NM_025391.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010215.mm.1 |
NM_025391.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010215.mm.1 |
NR_028367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010215.mm.1 |
NR_028367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010215.mm.1 |
TC0800001263.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010216.mm.1 |
NR_028367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010216.mm.1 |
NR_028367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010216.mm.1 |
TC0800001263.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010214.mm.1 |
NM_001164472.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010214.mm.1 |
NM_001164472.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010214.mm.1 |
NM_025391.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010214.mm.1 |
NM_025391.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010214.mm.1 |
NR_028367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010214.mm.1 |
NR_028367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010214.mm.1 |
TC0800001263.mm.1 |
MTA TC |