Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130868.2 | NP_001124340.1 | 1 - 2 | 152 | 59 | |
GenBank mRNA | BC035324.1 | - | 1 - 2 | 64 | 2811286 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |