Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
NM_001039390.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
NM_001039390.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
NM_001039391.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
NM_001039391.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
NM_001164053.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
NM_001164053.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
NM_001164055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
NM_001164055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
NM_001277096.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
NM_011106.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
NM_011106.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020359.mm.1 |
TC0200002494.mm.1 |
MTA TC |