Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rltpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033320.2 | NP_001028492.1 | 10 - 11 | 1472 | 71 | |
GenBank mRNA | AK031012.1 | - | 10 - 11 | 1471 | 71 | |
BC115830.1 | - | 2 - 3 | 92 | 71 | ||
HF678090.1 | - | 31 - 32 | 3103 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |