Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301173.1 | NP_001288102.1 | 6 - 7 | 1027 | 114 | |
NM_177791.3 | NP_808459.1 | 7 - 8 | 1096 | 114 | ||
GenBank mRNA | AK076644.1 | - | 7 - 8 | 1088 | 114 | |
BC092530.1 | - | 6 - 7 | 1027 | 6744085 | 114 | |
BC125431.1 | - | 7 - 8 | 1088 | 40129946 | 114 | |
BC125433.1 | - | 7 - 8 | 1088 | 40129948 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |