Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145439.2 | NP_663414.3 | 14 - 15 | 2030 | 61 | |
GenBank mRNA | AK077671.1 | - | 12 - 13 | 1775 | 61 | |
AK090179.1 | - | 14 - 15 | 1985 | 61 | ||
AY236497.1 | - | 14 - 15 | 1929 | 61 | ||
AY263158.1 | - | 14 - 15 | 1955 | 61 | ||
BC004840.1 | - | 14 - 15 | 1955 | 2648849 | 61 | |
BC005510.1 | - | 7 - 8 | 1000 | 61 | ||
BC006956.1 | - | 12 - 13 | 1740 | 61 | ||
BC013502.1 | - | 5 - 6 | 757 | 4223599 | 61 | |
BC058195.1 | - | 14 - 15 | 1955 | 5356348 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |