Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252578.1 | NP_001239507.1 | 3 - 4 | 847 | 76 | |
NM_001252579.1 | NP_001239508.1 | 3 - 4 | 693 | 76 | ||
NM_028072.5 | NP_082348.2 | 3 - 4 | 689 | 76 | ||
GenBank mRNA | AK028874.1 | - | 3 - 4 | 604 | 76 | |
AK034712.1 | - | 3 - 4 | 674 | 76 | ||
AK036685.1 | - | 3 - 4 | 625 | 76 | ||
AK129316.1 | - | 3 - 4 | 847 | 76 | ||
AK133336.1 | - | 663 | 76 | |||
AK165183.1 | - | 3 - 4 | 601 | 76 | ||
AY101177.1 | - | 2 - 3 | 508 | 76 | ||
BC062900.1 | - | 3 - 4 | 646 | 76 | ||
BC141086.1 | - | 2 - 3 | 442 | 9055698 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |