Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170704.2 | NP_733805.1 | 7 - 8 | 724 | 151 | |
GenBank mRNA | AJ401389.1 | - | 7 - 8 | 658 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |