Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011497.4 | NP_035627.1 | 2 - 3 | 217 | 114 | |
GenBank mRNA | AF007817.1 | - | 2 - 3 | 164 | 114 | |
BC014711.1 | - | 2 - 3 | 153 | 3968835 | 114 | |
CD560646.1 | - | 2 - 3 | 140 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |